Cole-Parmer® MP-800 Series Stuart Automatic Digital Melting Point Apparatus, Antylia Scientific
Supplier: Cole-Parmer
03013-02
75871-040EA 7837.63 USD
75871-040
Cole-Parmer® MP-800 Series Stuart Automatic Digital Melting Point Apparatus, Antylia Scientific
Melting Point Apparatus
Capture and display video image of the melt on touch-screen display or download to a PC.
  • Accurately identify the melting point with advanced digital image processing
  • Get up and running quickly - intuitive setup and navigation on large 7" color touch screen
  • Increase productivity - analyze three samples simultaneously
  • 8 GB storage holds approximately 300 melt files including videos
  • Set ramp rate from 0.1 to 20 °C in 0.1 °C increment
Advanced features with a familiar interface make this the ideal melting point apparatus for pharmaceutical, raw material, and quality control applications. Setup is easy using the intuitive menu system, based on an Android™ platform. Take measurements, view results, save programs, set up user sign-ins, and more. Unsure of the melt temperature? Use the rapid ramp mode to take a fast scan of the full temperature range. Watch the melt on the large 7" (17.8 cm) touch-screen display, or set the unit up and walk away and view a video of the melt later. Download videos and reports to a USB flash drive or use the optional printer for direct printing of results. Stored video results include a temperature stamp of the captured melt temperature. Storage compartment for unused capillaries includes a glass cutter to open capillaries that are closed on both ends. There is also an area to hold three prepared sample capillaries safely away from the heat source.
Ordering information: Supplied with glass capillaries and power cord.