Vydac 302IC, Anion Exclusion HPLC Column
Supplier: Avantor
302IC4.6
75830-792EA 336.32 USD
75830-792 75870-424 76398-194
Vydac 302IC, Anion Exclusion HPLC Column
Chromatography Columns
Vydac 302IC is a low capacity anion-exchange (quaternary amine) material based on a high purity 10 µm large pore size silica.
  • Used with standard HPLC systems for analysis of common ions
  • Widely used for environmental analysis
This column is well known for 'non-suppressed' ion chromatography applications.
Vydac is a registered trademark of W.R.Grace and Co.