Dilushaker III Digital Diluter
Supplier: Hardy Diagnostics
40DS21PRXT 40DEMRD6 40DEBPW6
75860-388EA 2904.02 USD
75860-388 75860-392 75860-406 75860-408 75860-410 75860-412 75860-414 75860-418
Dilushaker III Digital Diluter
Dilutors Volumetric Dilutors
For industries that perform serial dilutions, the Dilucup Elegance serial dilution system is the easiest, most cost effective answer in the field.
  • Adjustable vortex time from 1 to 15 seconds per dilution step
  • Removable tray for easy cleaning
  • Ramp with LED lights to guide the user
  • Adjustable direction of the ramp to suit both right- and left-handed users
  • Adjustable number of dilution steps per sample
  • Can be activated by foot switch or hands-free optical sensor
This system maximizes lab efficiency with its pre-filled, sterile and ready-to-use dilution cups.
You will also see measurable differences in the amount of storage space required for this product line. The compact and thoughtfully design packaging will save you on shipping and shelf space.
The Dilushaker III Digital has been developed for use with the Dilucup Elegance serial dilution cups.
These save time and standardize results. The lab will virtually eliminate results that vary from tech to tech. The Dilucup line also reduces the occurrence of repetitive motion injuries.
Ordering information: Power supply required (sold separately).
Delivery information: Includes extractible tray for 21 cups (7×3) or 42 cups (7×6).