VWR® Molasses for Fly Food
Supplier: VWR International
75860-374
75860-374EA 40.18 USD
75860-374
VWR® Molasses for Fly Food
Animal Supplies Animal Feed
Molasses for fly food.
  • Unsulfured Molasses
  • Contains pure molasses with no preservatives or additives
  • Ideal medium for Drosophila Food
This pure molasses is ideal for Drosophila food as it does not contain preservatives or additives. Unsulfured molasses offers the highest quality, and is extracted from the juice of sun-ripened cane.