Active Yeast, Azer Scientific
Supplier: VWR International
75860-348
75860-348EA 35.08 USD
75860-348
Active Yeast, Azer Scientific
Animal Supplies Animal Feed
Active Yeast for Fly Food
  • Prevents harmful molds from growing in a Drosophila culture
This active yeast is ideal for successful fly culture growth and is often used in conjunction with agar, molasses, and cornmeal.