Cornmeal, Azer Scientific
Supplier: VWR International
75860-346
75860-346EA 133.12 USD
75860-346
Cornmeal, Azer Scientific
Animal Supplies Animal Feed
Cornmeal for Fly Food
  • Yellow cornmeal
  • Often used with molasses, active yeast, and agar
  • For successful fly culture growth
This yellow cornmeal is used for successful fly culture growth. It is often used with molasses, active yeast, and agar.