PHCbi Cell IQ™ Dry Heat Sterilization CO₂ Incubator, PHC Corporation
Supplier: PHC Corporation
MCO-170AICUVDL-PA
75856-512EA 12384.1 USD
75856-512
PHCbi Cell IQ™ Dry Heat Sterilization CO₂ Incubator, PHC Corporation
Incubators CO₂ Incubators
Cell IQ™ Dry Heat Sterilization CO2 incubator provides optimal cultivation conditions with precise CO2 concentration. A unique dual heat system sterilizes the incubator interior.
  • Precise and repeatable
  • Dual heat sterilization
  • Antimicrobial interior
  • Optimum cell growth
  • Rapid CO2 recovery
  • Three-year manufacturer's parts and labor warranty
An integrated auxiliary sterilization system is utilized together with the incubator cultivation heaters to provide an effective, energy efficient sterilization process
InCu-saFe® and SafeCell™ UV function together to create contamination control against external microbes. InCu-saFe is comprised of a germicidal copper enriched stainless steel interior. SafeCell UV decontaminates circulated air and the humidity reservoir. A unique heating system and the dual filter infrared sensor precisely control temperature and CO2 levels, respectively. Sensor response accelerates CO2 recovery to setpoint without overshoot following frequent door openings.
A color LCD touchscreen allows full control and visibility of the incubator’s internal conditions, even with gloved hands. A USB port makes transferring data to a PC convenient. The incubator has fully rounded corners and shelf channels integrated into the chamber side walls for ease of cleaning and maintenance. Optimal results and reproducibility make the incubator ideal for tissue culture research, genomic expression, antibody production, transfection and transduction procedures.
Melamine foam insulation minimizes heat leakage to adjacent incubators and dramatically reduces elevated external surface temperatures. The incubator is engineered with a dual heat system. The primary heater maintains precise temperature control over a setpoint range from 5 °C above ambient to 50 °C. The secondary heater is activated when the dry heat sterilization cycle is initiated as both heaters combine to reach 180 °C.