AkroBins, Akro-Mils
Supplier: Akro-Mils
30210BLACK 30210GREEN 30220STONE 30224BLACK 30224BLUE 30224GREEN 30224RED 30224SCLAR 30224STONE 30224YELLO 30230BLACK 30230GREEN 30230SCLAR 30230STONE 30231CRY 30234BLACK 30234BLUE 30234GREEN 30234RED 30234SCLAR 30234STONE 30234YELLO 30235BLACK 30235RED 30235SCLAR 30235STONE 30235YELLO 30237BLACK 30237BLUE 30237GREEN 30237RED 30237SCLAR 30237STONE 30237YELLO 30239BLACK 30239GREEN 30239RED 30239SCLAR 30239STONE 30239YELLO 30240BLACK 30240GREEN 30240PURPL 30240SCLAR 30240STONE 30250BLACK 30250GREEN 30250SCLAR 30250STONE 30265GREEN 30265RED 30265SCLAR 30265STONE 30265YELLO 30270BLACK 30270BLUE 30270GREEN 30270RED 30270SCLAR 30270STONE 30270YELLO 30210SCLAR 30210STONE 30220BLACK 30220GREEN 30220SCLAR 30255BLACK 30255BLUE 30255GREEN 30255RED 30255SCLAR 30255STONE 30255YELLO 30260BLACK 30260BLUE 30260GREEN 30260RED 30260SCLAR 30260STONE 30260YELLO 30265BLACK 30265BLUE
75854-910CS 109.92 USD
75854-910 75854-912 75854-918 75854-922 75854-924 75854-926 75854-932 75854-934 75854-936 75854-938 75854-942 75854-944 75854-948 75854-950 75854-952 75854-954 75854-972 75854-974 75854-976 75854-982 75854-984 75854-986 75854-988 75854-990 75854-998 75855-000 75855-002 75855-004 75855-008 75855-010 75855-012 75855-014 75855-016 75855-018 75855-020 75855-022 75855-024 75855-030 75855-032 75855-034 75855-036 75855-038 75855-040 75855-042 75855-044 75855-046 75855-048 75855-058 75855-060 75855-064 75855-066 75855-068 75855-070 75855-072 75855-074 75855-076 75855-078 75855-080 75855-082 75855-084 75855-086 75855-088 75855-090 75855-092 75855-780 75855-782 75855-786 75855-788 75855-794 75855-796 75855-798 75855-800 75855-802 75855-804 75855-806 75855-808 75855-810 75855-812 75855-814 75855-816 75855-818 75856-038 75856-040 75856-042 75856-044
AkroBins, Akro-Mils
Boxes Storage Boxes
AkroBins are the industry-leading hanging and stacking plastic storage bin.
  • Anti-slide stop prevents stacked bins from shifting forward
  • Full-width hanger lip supports bin on hanging system
  • Made in the USA
Heavy-duty, industrial-grade polymer bins stack together or hang from Akro-Mils louvered racks and panels, rail racks, storage cabinets, and louvered carts for high-density storage. AkroBins are corrosion-proof and unaffected by weak acids and alkalis. Dividers, lids, and a wide range of labeling options are available separately.
Accessories information: Create multiple compartments within a single bin, and keep items separated with economical and sturdy length dividers. This provides additional storage options, to maximize space and efficiency.