VWR® Adhesive Sealing Films for PCR and Storage
Supplier: VWR International
75853-868 76449-250
75853-868PK 73.82 USD
75853-868 76449-250
VWR® Adhesive Sealing Films for PCR and Storage
Seals Microplate Seals
Polyester seals feature a strong adhesive to minimize evaporation during PCR and storage applications.
  • Ideal for PCR & storage
  • Certified DNase- and RNase- free
  • Temperature range is –40 to +120 °C
  • Non-pierceable
  • Minimizes evaporation
Removes easily without leaving adhesive residue.