Peroxigen ST Sterile, 6% Hydrogen Peroxide, Decon Labs
Supplier: Decon Labs
9716
75853-810CS 472.89 USD
75853-810
Peroxigen ST Sterile, 6% Hydrogen Peroxide, Decon Labs
Cleaning Agents Detergents and Surface Cleansers
Peroxigen ST is a sterile 6% (v/v) Hydrogen Peroxide solution made with water-for-injection (WFI).
  • Peroxigen ST is filtered to 0.2 microns
  • Bottled and double-bagged within a ISO CLASS 5 cleanroom
  • Gamma-irradiated to a sterility assurance level (SAL) of 10⁻⁶
This ready-to-use solution can be used to clean surfaces and equipment found in cleanroom areas within pharmaceutical, biotechnology, and medical device facilities, or anywhere the use of a sterile, hydrogen peroxide solution is required.
Each lot is tested for sterility via USP 14-day sterility test. A QR code can be found on each case of Peroxigen ST which is linked to a copy of the lot specific document, which details QC, irradiation, and sterility for that particular lot.