OIML Class 2 Individual Precision Weights, ASTM, Mettler Toledo
Supplier: Mettler Toledo
11123537 11123536 11123513 11123514
89422-920EA 4305 USD
89422-920 75852-744 75852-746 75852-880
OIML Class 2 Individual Precision Weights, ASTM, Mettler Toledo
Weights Calibration Weights
ASTM weights are designed to resist dust and moisture intrusion, offering long-term stability.
  • 316 stainless steel
  • Low magnetic permeability
  • High corrosion resistance
  • High gloss polish
All precision weights are protected in robust and easy to clean plastic boxes. FDA approved materials, including foam inserts, make them a perfect solution for regulated industries.