Standard Laboratory Use Sterilizer, 220 V, Yamato
Supplier: Yamato Scientific
SN-310C
75846-578EA 11353.93 USD
75846-578
Standard Laboratory Use Sterilizer, 220 V, Yamato
Sterilizers Autoclaves (Steam Sterilizers)
Features of the ergonomically designed, easy-to-use sterilizer include easy top loading and sample handling.
  • 220 V model
  • Wheels for Mobility Provided
  • Two Heaters
  • Lid Interlock Mechanism
  • Three Female Thread Ports for Sensors
  • Axial Fan Motor
The maximum sterilizing temperature goes up to 135 °C, and the operating temperature range is 45 to 135 °C. With simple setting and operation, this sterilizer manages ordinary sterilization, sterilization of culture mediums and liquids, and melting of culture mediums with its automatic high-pressure steam sterilization system.
Custom programs can be saved for repeated use, and the display indicates accumulated working and present time. Each sterilizer features an exhaust valve and a slow-release valve. Preheating and forced cooling functions are available. Operation modes include an instrument sterilization course, liquid sterilization course, sterilization and keep-warm course, key locking, and customer-programmed course. Safety modes include sensor failure detection, memory-error detection, automatic shut down during malfunction, and a pressure-release valve. The unit can save up to 20 error logs.
Ordering information: Mesh baskets provided. Additional mesh baskets available separately.
Caution: 220 V model. Corresponding model with voltage suitable for use in US (75846-576) is also available, please see separate product page.