Coliscreen, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z204
75846-318PK 390.36 USD
75846-318
Coliscreen, Hardy Diagnostics
Media for Microbiology Prepared Media for Microbiology
For the presumptive identification of Escherichia coli.
  • Uses the Indole and Glucuronidase tests
  • Presumptive identification of Escherichia coli
Packaging: 15/45 mm tube.