ALA Disks™ Delta-Aminolevulinic Acid, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z7081
75846-306PK 60.29 USD
75846-306
ALA Disks™ Delta-Aminolevulinic Acid, Hardy Diagnostics
Microbiology Test Systems Microorganism Tests
ALA Disks™, delta-aminolevulinic acid, for the differentiation and speciation of Haemophilus spp., a porphyrin test.
  • A useful aid for differentiating species of Haemophilus
  • Spring-loaded for smooth and reliable dispensing
  • Single metal disk dispenser
Each cartridge contains 50 disks that are spring-loaded for smooth and reliable dispensing from the single metal disk dispenser by BD BioSciences.
ALA Disks require the use of a handheld 366 nm ultra violet lamp.