BioClean™ DB Sterile Pouch-Style Facemasks, Nitritex
Supplier: Ansell Healthcare
BDBS
75845-808CS 661.54 USD
75845-808 75845-810
BioClean™ DB Sterile Pouch-Style Facemasks, Nitritex
Masks Cleanroom Masks
Latex-free facemask, with high BFE and PFE, ISO Class 4, and EU GMP Grade A compatibility.
  • High bacterial, particle, and viral filtration efficiency
  • Large breathing chamber
  • Ultrasonically sealed edges
  • Fully enclosed malleable nose-band
The BioClean™ DB Pouch-style sterile facemask features a large breathing chamber for increased wearer comfort and a malleable nose-band for a good fit. Made from cleanroom compatible materials with ultrasonically sealed edges to reduce the risk of contamination entering a controlled environment.
Facemask BDBS-G has additional neck protection to cover the gap between mask and body protection.
Certifications: Manufactured in a facility holding ISO 9001:2008.
Processed in an NEBB certified ISO Class 4 cleanroom environment.
Sampling and inspection in accordance with ISO 2859-3:1991.