Combination Refrigerator/Freezers, Marvel Scientific
Supplier: Marvel Scientific
MS24RFS4RW
75845-464EA 9237.92 USD
75845-464
Combination Refrigerator/Freezers, Marvel Scientific
Refrigerators
Powerful compressor for fast pull-down time and temperature recovery after door opening.
  • Three cantilevered full-depth perforated metal shelves
  • MaxStore™ roller-glide clear utility bin provides maximum volume to effectively store large, bulky or loose items
  • Internal side-mounted 1 cu. ft. freezer with self-closing door
  • “Box Style” door for easy mounting of inventory management system hardware
  • Door lock to secure contents
  • Cam-assisted door close hinge system
Accurate, externally mounted MicroSentry II™ electronic temperature control within ±2 °C of set point with settings between +1° and +6 °C