Master-Mite® Heat Gun, Master Appliance
Supplier: Master Appliance
10008
75845-352EA 256.91 USD
75845-352
Master-Mite® Heat Gun, Master Appliance
Heaters Hot Air Dryers
A continuous duty bench-top heating system with 650°F element, stand and heat shrink attachment.
  • Quiet running motor, eliminates noisy production areas
  • Three optional easy-to-change plug-in heating elements available: 500°, 650° and 800°F
Complete system comes standard with 650°F heating element, adjustable heat gun stand and heat shrink attachment. Ideal heat and air flow (3.8 CFM), performs bench top heating jobs without excess air flow, won’t blow components around work station. Adjustable heat gun is used to activate adhesive, cure epoxies, dry components, melt solder performs, remove decals, shrink tubing and more.