Open-Head UN Rated Poly Drum, With Metal Lever-Lock Ring, Eagle Mfg
Supplier: Eagle Manufacturing
1656MB
75845-054EA 317.94 USD
75845-054
Open-Head UN Rated Poly Drum, With Metal Lever-Lock Ring, Eagle Mfg
Barrels, Kegs, Drums
Drum is constructed of blue blow-molded high-density polyethylene (HDPE) with UV inhibitors and has a white lid with a metal lever-lock band closure.
  • Durable
  • Lightweight
  • Chemical and weather resistant
Ideal for safe storage, handling, and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal, and incineration.
Certifications: Complies to DOT 46, CFR 173.12, and UN DOT Rating 1H2/X200/S, 1H2/Y250/S.