CHIR99021 ≥95%, crystalline solid

Supplier: BIOGEMS INTERNATIONAL INC.

2520691-1MG 2520691-10MG
75844-608EA 52.25 USD
75844-608 75844-610
CHIR99021 ≥95%, crystalline solid
CHIR99021

CHIR 99021 is an aminopyrimidine derivate that is a selective and potent inhibitor of glycogen synthase kinase 3 (GSK-3)


This has been used to maintain human and mouse stem cell lines. In combination with other reagents involves in chemical reprogram of somatic cells into stem cells.


Soluble in organic solvents such as DMSO

Formula: C₂₂H₁₈Cl₂N₈
Storage Temperature: Freezer
MDL Number: MFCD11846251
CAS Number: 252917-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR