Anti-TIM3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: F38-2E2]

Supplier: BIOGEMS INTERNATIONAL INC.
78211-80-25
75844-516EA 249.88 USD
75844-516
Anti-TIM3 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: F38-2E2]
Antibodies
The F38-2E2 monoclonal antibody specifically reacts with human T cell immunoglobulin and mucin domain containing protein (TIM3), a transmembrane molecule found on monocytes, macrophages, dendritic cells, and activated T cell types such as Treg, Th1, and Th17. TIM3 can exist in soluble and membrane bound forms and the relevant ligand for the protein is Galectin-9. It plays a role in the creation of immunological tolerances, the secretion of Th1 and Th17 related cytokines, and HIV infection.

Type: Primary
Antigen: TIM3
Clonality: Monoclonal
Clone: F38-2E2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR