CD339(Jagged 1) MAB 25ug
Supplier: Biogems
CD339(Jagged 1) Monoclonal Antibody, Clone: HMJ1-29, Host: Armenian Hamster, Species reactivity: Mouse, Isotype: IgG, Conjugate: PE, Formulation: Phosphate-buffered aqueous solution,