Anti-JAG1 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: HMJ1-29]

Supplier: Biogems
41412-60-100
75844-504EA 716.42 USD
75844-504
Anti-JAG1 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: HMJ1-29]
Antibodies
The HMJ1-29 monoclonal antibody reacts with mouse and rat CD339 (Jagged 1), a type 1 transmembrane Notch receptor ligand. It is expressed by macrophages and certain stromal, epithelial, dendritic, and thymic lymphoid cells. It has been reported to be play a role in Treg induction, Th2 polarization, neurogenesis, and cardiovascular development.

Type: Primary
Antigen: JAG1
Clonality: Monoclonal
Clone: HMJ1-29
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR