Anti-PD-1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MIH4]

Supplier: Biogems
31811-50-100
75844-496EA 562.67 USD
75844-496
Anti-PD-1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: MIH4]
Antibodies
The MIH4 monoclonal antibody specifically reacts with human Programmed death-1 (PD-1 or CD279), a 50-55 kDA glycoprotein. It is expressed on mainly on activated B, T, and myeloid cells. Within the cytoplasmic region, PD-1 contains an Immunoreceptor tyrosine-based inhibitory motif (ITIM) and seems to regulate peripheral tolerance. The lack or mutation of CD279 is linked to autoimmune disorders.

Type: Primary
Antigen: PD-1
Clonality: Monoclonal
Clone: MIH4
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR