Anti-IL-7RA Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: A7R34]

Supplier: Biogems
19212-77-100
75844-480EA 374.21 USD
75844-480
Anti-IL-7RA Rat Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: A7R34]
Antibodies
The A7R34 monoclonal antibody specifically reacts with the mouse CD127, the α subunit of the IL-7 receptor, expressed by immature B lymphocytes in the bone marrow, CD4-/CD8-, CD4+, and CD8+ thymocytes, and by mature T lymphocytes at low levels. Mature T cells express CD117 at low levels in the periphery.The A7R34 antibody prevents the interaction between IL-7 and its receptor and the binding of mAb SB/199, which also recognizes mouse CD127.

Type: Primary
Antigen: IL-7RA
Clonality: Monoclonal
Clone: A7R34
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR