Anti-IL-7Ra Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: A7R34]

Supplier: Biogems
19212-50-25
75844-468EA 51.63 USD
75844-468
Anti-IL-7Ra Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: A7R34]
Antibodies
The A7R34 monoclonal antibody specifically reacts with the mouse CD127, the α subunit of the IL-7 receptor, expressed by immature B lymphocytes in the bone marrow, CD4-/CD8-, CD4+, and CD8+ thymocytes, and by mature T lymphocytes at low levels. Mature T cells express CD117 at low levels in the periphery.The A7R34 antibody prevents the interaction between IL-7 and its receptor and the binding of mAb SB/199, which also recognizes mouse CD127.

Type: Primary
Antigen: IL-7Ra
Clonality: Monoclonal
Clone: A7R34
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR