Anti-IL-7RA Rat Monoclonal Antibody (BG Violet 450) [clone: A7R34]

Supplier: Biogems
19212-40-25
75844-464EA 174.21 USD
75844-464
Anti-IL-7RA Rat Monoclonal Antibody (BG Violet 450) [clone: A7R34]
Antibodies
The A7R34 monoclonal antibody specifically reacts with the mouse CD127, the α subunit of the IL-7 receptor, expressed by immature B lymphocytes in the bone marrow, CD4-/CD8-, CD4+, and CD8+ thymocytes, and by mature T lymphocytes at low levels. Mature T cells express CD117 at low levels in the periphery.The A7R34 antibody prevents the interaction between IL-7 and its receptor and the binding of mAb SB/199, which also recognizes mouse CD127.BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Primary
Antigen: IL-7RA
Clonality: Monoclonal
Clone: A7R34
Conjugation: BG Violet 450
Epitope:
Host: Rat
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR