CD127(IL-7RA) MAB 100ug
Supplier: Biogems
CD127(IL-7RA) Monoclonal Antibody, Clone: A7R34, Host: Rat, Species reactivity: Mouse, Isotype: IgG2a, Kappa, Conjugate: Purified, Formulation: Phosphate-buffered aqueous solution,