CD117(c-Kit) MAB 25ug
Supplier: Biogems
CD117(c-Kit) Monoclonal Antibody, Clone: ACK2, Host: Rat, Species reactivity: Mouse, Isotype: IgG2b, Kappa, Conjugate: APC, Formulation: Phosphate-buffered aqueous solution,