Anti-CD117 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: ACK2]

Supplier: Biogems
19112-80-100
75844-446EA 206.45 USD
75844-446
Anti-CD117 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: ACK2]
Antibodies
The ACK2 monoclonal antibody specifically reacts with mouse CD117 (c-Kit receptor), a 145 kDa transmembrane tyrosine-kinase receptor encoded by the Kit gene. The c-Kit receptor, also known as stem cell factor receptor, is expressed on hematopoietic progenitor cells in adult bone marrow, in progenitors of erythroid and myeloid lineages, and precursors of B and T cells. CD117 enhances the proliferation and the differentiation of the hematopoietic progenitor cells and seems to enhance the development of T cells, as the c-Kit receptor and its ligand are expressed by the thymus.

Type: Primary
Antigen: CD117
Clonality: Monoclonal
Clone: ACK2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR