Anti-CD117 (c-Kit) (ACK2) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: ACK2]

Supplier: Biogems
19112-60-25
75844-440EA 64.53 USD
75844-440
Anti-CD117 (c-Kit) (ACK2) Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: ACK2]
Antibodies
The ACK2 monoclonal antibody specifically reacts with mouse CD117 (c-Kit receptor), a 145 kDa transmembrane tyrosine-kinase receptor encoded by the Kit gene. The c-Kit receptor, also known as stem cell factor receptor, is expressed on hematopoietic progenitor cells in adult bone marrow, in progenitors of erythroid and myeloid lineages, and precursors of B and T cells. CD117 enhances the proliferation and the differentiation of the hematopoietic progenitor cells and seems to enhance the development of T cells, as the c-Kit receptor and its ligand are expressed by the thymus.

Type: Primary
Antigen: CD117 (c-Kit) (ACK2)
Clonality: Monoclonal
Clone: ACK2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR