Anti-CD146 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P1H12]

Supplier: Biogems
18512-60-25
75844-414EA 205.56 USD
75844-414
Anti-CD146 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: P1H12]
Antibodies
The P1H12 monoclonal antibody reacts with CD146, 118kDA transmembrane glycoprotein that is also called the melanoma cell adhesion molecule (Mel-CAM), MUC18, Endo-CAM, and S-Endo. It is a prominently glycosylated protein with carbohydrates comprising the majority of its mass and it is expressed on melanoma, stromal, endothelial, and bone marrow fibroblasts. The P1H12 antibody cross reacts with human, mouse, canine, rabbit to the exclusion of rat protein.

Type: Primary
Antigen: CD146
Clonality: Monoclonal
Clone: P1H12
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR