Anti-CD122 Rat Monoclonal Antibody [clone: TM-beta1]

Supplier: Biogems
14312-25-500
75844-388EA 716.42 USD
75844-388
Anti-CD122 Rat Monoclonal Antibody [clone: TM-beta1]
Antibodies
The TM-beta1 monoclonal antibody specifically reacts with mouse CD122, a beta chain shared by the IL-2 and IL-15 receptors. It is expressed by B cells, T cells, and NK cells and can be used as a marker for NK-cell progenitors. CD122 forms complexes involved in immunoregulation and signal transduction. The TM-beta1 antibody is reported to block the binding of IL-2 and IL-15 to the IL-2 beta receptor.

Type: Primary
Antigen: CD122
Clonality: Monoclonal
Clone: TM-beta1
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2b
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR