Anti-CD161 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: HP-3G10]

Supplier: Biogems
13711-70-25
75844-378EA 167.76 USD
75844-378
Anti-CD161 Mouse Monoclonal Antibody (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®) [clone: HP-3G10]
Antibodies
The HP-3G10 monoclonal antibody specifically binds to an 80 kDa homodimer type II membrane glycoprotein from the C-type lectin superfamily, known as the human CD161 or NKR-P1A. CD161 is expressed on most natural killer cells, subsets of CD4+ and CD8+ T lymphocytes, γδ TCR T lymphocytes, a subset of CD3+ thymocytes, and especially on CD45RO+ T lymphocytes. Reports indicate that it may serve as a specific receptor for some natural killer cell targets and a possible stimulatory role.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR