Anti-CD161 Mouse Monoclonal Antibody (BG Violet 450) [clone: HP-3G10]

Supplier: Biogems
13711-40-25
75844-366EA 141.82 USD
75844-366
Anti-CD161 Mouse Monoclonal Antibody (BG Violet 450) [clone: HP-3G10]
Antibodies
The HP-3G10 monoclonal antibody specifically binds to an 80 kDa homodimer type II membrane glycoprotein from the C-type lectin superfamily, known as the human CD161 or NKR-P1A. CD161 is expressed on most natural killer cells, subsets of CD4+ and CD8+ T lymphocytes, γδ TCR T lymphocytes, a subset of CD3+ thymocytes, and especially on CD45RO+ T lymphocytes. Reports indicate that it may serve as a specific receptor for some natural killer cell targets and a possible stimulatory role. BG Violet 450 conjugate is an alternative to the Pacific Blue, eFluor 450, or BD Horizon V450 dyes. It is excited by the violet (405 nm) laser, with a peak emission of 450nm.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: BG Violet 450
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR