Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: HIB19]

Supplier: Biogems
11211-77-100
75844-358EA 283.89 USD
75844-358
Anti-CD19 Mouse Monoclonal Antibody (PE (Phycoerythrin)-Cy7®) [clone: HIB19]
Antibodies
The HIB19 monoclonal antibody reacts with a human 95 kDa transmembrane glycoprotein known as CD19, which is expressed by B lymphocytes during all the developmental stages, except for the terminally differentiated plasma cells. CD19 is also expressed on follicular dendritic cells, and seems to ensure the regulation of B lymphocytes proliferation. CD19, CD21, CD81, MHC class II, and Leu13 can bind together and form a complex which associates with the B cell receptor (BCR) on the surface of B lymphocytes and facilitates the signal transduction.

Type: Primary
Antigen: CD19
Clonality: Monoclonal
Clone: HIB19
Conjugation: PE (Phycoerythrin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR