Anti-CD48 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: HM48-1]

Supplier: Biogems
10512-60-100
75844-340EA 447.86 USD
75844-340
Anti-CD48 Armenian Hamster Monoclonal Antibody (PE (Phycoerythrin)) [clone: HM48-1]
Antibodies
The HM48-1 monoclonal antibody specifically reacts with mouse CD48, a GPI-anchored molecule in the Ig superfamily. CD48 was previously known as BCM1 in mice, Blast-1 in humans, and OX-45 in rats. Its ligands are CD2 and CD244 and it is expressed on leukocytes to the exclusion of non-hematopoietic cells. The utilization of the HM48-1 antibody has been reported to prolong cardiac allografts survival in in vivo studies. The antibody blocks the interaction of CD2 and CD244, inhibits CTL priming, and blocks the binding of soluble CD2 to CD48 expressing cells.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: HM48-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Armenian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR