CD48 MAB 500ug
Supplier: Biogems
CD48 Monoclonal Antibody, Clone: 156-4H9, Host: Mouse, Species: Human, Isotype: IgG1, Conjugate: SAFIRE Purified, Formulation: Phosphate-buffered aqueous solution, ph7.2, Expressed on leukocytes to the exclusion of non-hematopoietic cells, Application: FC, FA, Size: 500ug