Anti-CD48 Mouse Monoclonal Antibody [clone: 156-4H9]

Supplier: Biogems
10511-25-50
75844-330EA 243.02 USD
75844-330
Anti-CD48 Mouse Monoclonal Antibody [clone: 156-4H9]
Antibodies
The 156-4H9 monoclonal antibody specifically reacts with human CD48, a GPI-linked glycoprotein in the Ig superfamily. CD48 was previously known as BCM1 in mice, Blast-1 in humans, and OX-45 in rats. Its ligands are CD2 and CD244 and it is expressed on leukocytes to the exclusion of non-hematopoietic cells and its expression increases in B cells upon activation.

Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 156-4H9
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR