Anti-CD38 Mouse Monoclonal Antibody [clone: 90]

Supplier: Biogems
10412-25-500
75844-328EA 640.33 USD
75844-328
Anti-CD38 Mouse Monoclonal Antibody [clone: 90]
Antibodies
The 90 monoclonal antibody specifically bind to mouse CD38, a type II transmembrane glycoprotein. CD38 is expressed on activated T cells, plasma cells, monocytes, and macrophages. In the mouse as opposed to the human, CD38 expression is downregulated on mouse germinal center B cells and plasma cells. CD38 displays cyclase and hydrolase enzymatic activity and is involved in lymphocyte adhesion, activation, and cell signaling. Its ligand is CD31.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: 90
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR