Anti-CD38 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIT2]

Supplier: BIOGEMS INTERNATIONAL INC.
10411-80-100
75844-322EA 309.69 USD
75844-322
Anti-CD38 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HIT2]
Antibodies
The HIT2 monoclonal antibody specifically bind to human CD38, a 45kda type II transmembrane glycoprotein. CD38 is expressed on activated T cells, plasma cells, monocytes, macrophages, dendritic cells, and thymocytes. CD38 displays cyclase and hydrolase enzymatic activity and is involved in lymphocyte adhesion, activation, and cell signaling. Its ligand is CD31.

Type: Primary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR