Anti-CD28 Mouse Monoclonal Antibody [clone: JJ319]

Supplier: Biogems
10313-25-500
75844-306EA 660.09 USD
75844-306
Anti-CD28 Mouse Monoclonal Antibody [clone: JJ319]
Antibodies
The JJ319 monoclonal antibody specifically reacts with rat CD28, a 44 kDa glycoprotein member of the immunoglobulin superfamily. CD28 is expressed on alpha/beta TCR+ T cells, some gamma/delta TCR+ T cells and a subset of NK cells. CD28 is a costimulatory receptor required for T cell activation. Its ligands are CD80 (B7-1) and CD86 (B7-2).

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: JJ319
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR