Anti-CD28 Golden Syrian Hamster Monoclonal Antibody [clone: 37.51]

Supplier: Biogems
10312-20-100
75844-294EA 109.69 USD
75844-294
Anti-CD28 Golden Syrian Hamster Monoclonal Antibody [clone: 37.51]
Antibodies
The 37.51 monoclonal antibody specifically reacts with mouse CD28 molecule, a 45 kDa homodimer. It is expressed on most thymocytes, at low density on most of the CD4+ and CD8+ peripheral T lymphocytes, and on the natural killer cells. It is additionally found on activated splenocytes, mast cells, and thymocytes. CD 28 is a costimulator of T lymphocytes and a ligand for CD80 (B7-1), and CD86 (B7-2). The CD28 signaling stimulates the expression of IL-2 and IL-2 receptor and the cytotoxicity of CD3-activated T lymphocytes.The 37.51 antibody stimulates the proliferation of T lymphocytes and natural killer cells and their respective cytokine production.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: 37.51
Conjugation:
Epitope:
Host: Golden Syrian Hamster
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR