Anti-CD28 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CD28.2]

Supplier: Biogems
10311-80-25
75844-292EA 135.51 USD
75844-292
Anti-CD28 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CD28.2]
Antibodies
The CD28.2 monoclonal antibody specifically binds with the human 44 kDa homodimeric trans-membrane glycoprotein CD28, expressed on the surface of most mature T lymphocytes, plasma cells, and thymocytes. CD28 is a ligand for B7-1 (CD80) and B7-2 (CD86), a co-stimulator of T lymphocytes, and enhances the interaction between T and B lymphocytes. It has been reported that the T lymphocytes stimulation to produce IL-2 depends on the monoclonal antibody involved, which suggests that the CD28 molecule presents some subregions with distinct functions. The CD28.2 antibody induces Ca2+ influx in Jurkat T lymphocytes. Other studies have shown that CD28 is involved in the signal transduction.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR