CD8A MAB 25ug
Supplier: Biogems
CD8A Monoclonal Antibody, Clone: 53-6.7, Host: Rat, Species reactivity: Mouse, Isotype: IgG2a, kappa, Conjugate: FITC, Formulation: Phosphate-buffered aqueous solution, less than or equal to 0.09% Sodium azide, may contain carrier protein/stabilizer, ph7.2, Application: FC, Size: 25ug