Anti-NCAM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CMSSB]

Supplier: Biogems
08611-80-25
75844-212EA 249.6 USD
75844-212
Anti-NCAM Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: CMSSB]
Antibodies
The CMSSB monoclonal antibody specifically reacts with human CD56, or Neural Cell Adhesion molecule (NCAM). CD56 is a 140 kDa transmembrane glycoprotein also known as Leu-19 or NKH1. It is expressed on NK and NKT cells and has a role in cellular adhesion.

Type: Primary
Antigen: NCAM
Clonality: Monoclonal
Clone: CMSSB
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR