Anti-NCAM Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CMSSB]

Supplier: BIOGEMS INTERNATIONAL INC.
08611-60-25
75844-208EA 224.07 USD
75844-208
Anti-NCAM Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CMSSB]
Antibodies
The CMSSB monoclonal antibody specifically reacts with human CD56, or Neural Cell Adhesion molecule (NCAM). CD56 is a 140 kDa transmembrane glycoprotein also known as Leu-19 or NKH1. It is expressed on NK and NKT cells and has a role in cellular adhesion.

Type: Primary
Antigen: NCAM
Clonality: Monoclonal
Clone: CMSSB
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR