CD45.2 MAB 100ug
Supplier: Biogems
CD45.2 Monoclonal Antibody, Clone: 104, Host: Mouse, Species: Mouse, Isotype: IgG2a, kappa, Conjugate: PerCP-Cyanine5.5, Formulation: Phosphate-buffered aqueous solution,