Anti-CD45RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI100]

Supplier: BIOGEMS INTERNATIONAL INC.
07121-80-100
75844-122EA 309.69 USD
75844-122
Anti-CD45RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: HI100]
Antibodies
The HI100 monoclonal antibody specifically reacts with human CD45RA, the 220 kDa isoform of the human leukocyte common antigen (LCA) found on 40-50% of the peripheral CD4+ T lymphocytes, half of the peripheral CD8+ T lymphocytes and some of the monocytes and B lymphocytes. The CD45RA antigen is expressed by naïve and activated T lymphocytes. The HI100 monoclonal antibody is used as a phenotypic marker to discriminate T lymphocytes subsets.

Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: HI100
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR