Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin)-Cy7®) [clone: HI30]

Supplier: BIOGEMS INTERNATIONAL INC.
07111-87-100
75844-102EA 309.69 USD
75844-102
Anti-PTPRC Mouse Monoclonal Antibody (APC (Allophycocyanin)-Cy7®) [clone: HI30]
Antibodies
The HI30 monoclonal antibody specifically reacts with the 180 kDa, 190 kDa, 205 kDa, and 220 kDa isoforms of the human leukocyte common antigen (LCA) CD45. It is expressed on lymphocytes, granulocytes, monocytes, thymocytes, and eosinophils, but not on mature erythrocytes, platelets, mature erythroid cells of bone marrow, and non-hematopoietic tissues. CD45 is essential for T cell activation and the tyrosine phosphatase activity of its intracellular region is integral for signal transduction.

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: HI30
Conjugation: APC (Allophycocyanin)-Cy7®
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR