Anti-ICAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 15.2]

Supplier: BIOGEMS INTERNATIONAL INC.
06611-60-25
75844-064EA 122.58 USD
75844-064
Anti-ICAM1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 15.2]
Antibodies
The 15.2 antibody reacts with human CD54, also known as ICAM-1 (Intercellular Adhesion Molecule 1), a 90-110 kDa cell surface glycoprotein that is inducibly expressed on both immune and endothelial cells. As its name implies, ICAM-1 participates in cell-cell adhesion between leukocytes and endothelial cells, facilitating leukocyte recruitment and transmigration at sites of inflammation. The ligands for ICAM-1 are also expressed on leukocyte and endothelial cells, and include Mac-1, fibrinogen, and a member of the integrin protein family, LFA-1 (CD11a).The 15.2 antibody may be used for analysis of ICAM-1 expression in human cells and tissues, and is reported to be cross-reactive with porcine ICAM-1.

Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: 15.2
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR